Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021022.3 | NP_066302.2 | 25 | 3361 | ||
GenBank mRNA | AF133903.1 | - | 25 | 3332 | ||
AK144553.1 | - | 26 | 3436 |