Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021279.4 | NP_067254.1 | 4 | 1679 | ||
GenBank mRNA | AK141669.1 | - | 4 | 1534 | ||
BC005449.1 | - | 4 | 1668 | 3493095 | ||
M11943.1 | - | 4 | 1531 |