Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021297.2 | NP_067272.1 | 1, 2 | 119 | ||
GenBank mRNA | AF095353.1 | - | 1, 2 | 305 | ||
AF110133.1 | - | 1, 2 | 119 | |||
AF185285.1 | - | 1, 2 | 98 | |||
AK014533.1 | - | 320 | ||||
AK030348.1 | - | 1, 2 | 319 | |||
AK154297.1 | - | 1, 2 | 212 | |||
AK170566.1 | - | 1, 2 | 170 | |||
BC029856.1 | - | 1, 2 | 331 | 3493732 | ||
JX878358.1 | - | 1, 2 | 113 | |||
JX878359.1 | - | 1, 2 | 113 |