Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038604.1 | NP_001033693.1 | 7 | 803 | ||
NM_021364.2 | NP_067339.1 | 6 | 728 | |||
GenBank mRNA | AF139769.1 | - | 6 | 698 | ||
AK036697.1 | - | 6 | 694 | |||
AK046600.1 | - | 6 | 712 | |||
AK137482.1 | - | 6 | 729 | |||
AK155409.1 | - | 4 | 548 | |||
BC104364.1 | - | 7 | 762 | 40049817 | ||
BC104365.1 | - | 7 | 762 | 40049818 |