Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098668.3 | NP_001092138.1 | 6 | 1777 | ||
NM_001320813.1 | NP_001307742.1 | 6 | 1780 | |||
NM_001320814.1 | NP_001307743.1 | 5 | 1846 | |||
XM_005270128.3 | XP_005270185.1 | 6 | 1758 | |||
XM_005270132.3 | XP_005270189.1 | 5 | 1745 | |||
XM_011540124.1 | XP_011538426.1 | 6 | 1873 | |||
XM_011540125.1 | XP_011538427.1 | 6 | 1816 | |||
XM_017016608.1 | XP_016872097.1 | 5 | 1669 | |||
GenBank mRNA | HQ021421.1 | - | 6 | 1726 | ||
HQ021422.2 | - | 6 | 1726 | |||
HQ021423.2 | - | 6 | 1726 | |||
HQ021424.1 | - | 6 | 1726 | |||
HQ021425.2 | - | 6 | 1726 | |||
HQ021426.1 | - | 6 | 1723 | |||
HQ021427.1 | - | 6 | 1723 | |||
HQ021428.2 | - | 6 | 1723 | |||
HQ021429.2 | - | 6 | 1723 | |||
HQ021430.1 | - | 6 | 1723 | |||
HQ021431.2 | - | 6 | 1723 | |||
HQ021432.1 | - | 6 | 1726 |