Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277957.1 | NP_001264886.1 | 5 | 637 | ||
NM_001277958.1 | NP_001264887.1 | 5 | 626 | |||
NM_001277959.1 | NP_001264888.1 | 5 | 605 | |||
NM_021466.3 | NP_067441.1 | 5 | 637 | |||
GenBank mRNA | AK040747.1 | - | 5 | 542 | ||
AK132811.1 | - | 5 | 591 | |||
AK133426.1 | - | 5 | 627 | |||
BC061106.1 | - | 5 | 522 | 6595990 | ||
BC138425.1 | - | 5 | 503 | 8861445 | ||
Y09972.1 | - | 5 | 539 |