Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021886.1 | NP_068686.1 | 7 | 546 | ||
GenBank mRNA | AB017634.1 | - | 7 | 546 | ||
AK006208.1 | - | 7 | 530 | |||
AK010533.1 | - | 4 | 608 | |||
AK011264.1 | - | 7 | 531 | |||
AK011738.1 | - | 7 | 531 | |||
AK012907.1 | - | 7 | 491 | |||
BC025084.1 | - | 7 | 497 | 3501244 |