Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022813.3 | NP_073724.1 | 8 | 866 | ||
GenBank mRNA | AF295402.1 | - | 8 | 870 | ||
AK003080.1 | - | 8 | 866 | |||
AK161002.1 | - | 8 | 866 | |||
AK171766.1 | - | 8 | 857 | |||
BC003452.1 | - | 8 | 840 | |||
BC014751.1 | - | 8 | 843 | 4235556 |