Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022813.3 | NP_073724.1 | 9 | 1928 | ||
GenBank mRNA | AK161002.1 | - | 9 | 1927 | ||
AK171766.1 | - | 9 | 1917 | |||
BC014751.1 | - | 9 | 1906 | 4235556 | ||
CA979044.1 | - | 1 | 737 | |||
CX217627.1 | - | 1 | 14 |