Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sigirr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023059.3 | NP_075546.2 | 9 | 1567 | ||
GenBank mRNA | AF113795.1 | - | 8 | 1084 | ||
AF239957.1 | - | 8 | 1078 | |||
AK133518.1 | - | 9 | 1192 | |||
AK145889.1 | - | 9 | 1570 | |||
BC010806.1 | - | 8 | 1089 | 4240425 | ||
BC094069.1 | - | 9 | 1173 | 4974962 |