Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023314.3 | NP_075803.2 | 7 | 802 | ||
GenBank mRNA | AK018516.1 | - | 7 | 800 | ||
AK076067.1 | - | 5 | 517 |