Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164406.1 | NP_001157878.1 | 5 | 715 | ||
NM_025291.3 | NP_079567.2 | 5 | 818 | |||
GenBank mRNA | AK007302.1 | - | 5 | 804 | ||
AK010375.1 | - | 5 | 804 | |||
AK154232.1 | - | 5 | 788 | |||
AK180031.1 | - | 2 | 341 | |||
BC026480.1 | - | 3 | 597 | |||
BC048362.1 | - | 5 | 790 | 2646489 | ||
BQ748094.1 | - | 3 | 486 |