Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Natd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025294.5 | NP_079570.1 | 3 | 2983 | ||
GenBank mRNA | AK004720.2 | - | 3 | 2982 | ||
AK080453.1 | - | 3 | 2984 | |||
AK089179.1 | - | 3 | 2986 | |||
AK181642.1 | - | 1 | 317 | |||
AK182434.1 | - | 1 | 46 | |||
AK190677.1 | - | 1 | 59 | |||
AK192155.1 | - | 1 | 37 | |||
AK198941.1 | - | 1 | 319 | |||
AK200627.1 | - | 1 | 318 | |||
AK204811.1 | - | 1 | 18 | |||
AK209186.1 | - | 1 | 317 | |||
AK209571.1 | - | 1 | 318 | |||
AK215914.1 | - | 1 | 27 | |||
AK218977.1 | - | 1 | 318 | |||
AK219404.1 | - | 1 | 30 | |||
BC034332.1 | - | 3 | 2932 | 5025968 | ||
BC038816.1 | - | 3 | 2974 | 3490657 |