Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177658.1 | NP_001171129.1 | 3 | 323 | ||
NM_025300.4 | NP_079576.1 | 3 | 323 | |||
GenBank mRNA | AK003367.1 | - | 3 | 307 | ||
AK010474.1 | - | 3 | 283 | |||
AK010604.1 | - | 3 | 316 | |||
AK011775.1 | - | 2 | 143 | |||
AK017804.1 | - | 3 | 300 | |||
AK017820.1 | - | 3 | 302 | |||
AK017824.1 | - | 3 | 286 | |||
AK019175.1 | - | 3 | 296 | |||
AK019244.1 | - | 3 | 283 | |||
AK019249.1 | - | 3 | 283 | |||
AK150880.1 | - | 3 | 324 | |||
AK154840.1 | - | 1 | 1847 | |||
AK160660.1 | - | 3 | 281 | |||
BC021304.1 | - | 3 | 319 | 3498041 | ||
BC027233.1 | - | 3 | 308 | 3498065 | ||
BC068230.1 | - | 3 | 306 | |||
BY083200.1 | - | 3 | 338 |