Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025407.2 | NP_079683.2 | 4, 5 | 496 | ||
GenBank mRNA | AK002292.1 | - | 4, 5 | 437 | ||
AK004031.1 | - | 4, 5 | 442 | |||
AK010553.1 | - | 4, 5 | 436 | |||
AK013128.1 | - | 4, 5 | 436 | |||
AK151764.1 | - | 4, 5 | 438 | |||
AK160337.1 | - | 4, 5 | 443 | |||
AK167911.1 | - | 4, 5 | 436 | |||
AK168218.1 | - | 4, 5 | 438 | |||
AK177461.1 | - | 3, 4 | 289 | |||
AK183410.1 | - | 3, 4 | 289 | |||
AK184915.1 | - | 3, 4 | 289 | |||
AK185377.1 | - | 3, 4 | 289 | |||
BC030064.1 | - | 4, 5 | 433 | 1400271 |