Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030239.2 | NP_084515.2 | 4 | 536 | ||
GenBank mRNA | AF324242.1 | - | 4 | 488 | ||
AK030831.1 | - | 4 | 538 | |||
AK037614.1 | - | 4 | 452 | |||
AK051880.1 | - | 4 | 502 | |||
AK156626.1 | - | 4 | 504 | |||
BC138772.1 | - | 4 | 517 | 8861794 | ||
BC138773.1 | - | 4 | 516 | 8861795 |