Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPK1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006952.3 | NP_008883.2 | 5 | 480 | ||
GenBank mRNA | AB002155.1 | - | 5 | 438 | ||
AB015234.1 | - | 4 | 391 | |||
AF042331.1 | - | 4 | 401 | |||
AF082888.1 | - | 4 | 361 | |||
AK225868.1 | - | 4 | 371 | |||
AK303924.1 | - | 4 | 444 | |||
AK313787.1 | - | 5 | 502 | |||
AL709739.1 | - | 5 | 481 | |||
BC063568.1 | - | 4 | 428 | 3861427 | ||
CR457084.1 | - | 4 | 391 |