Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083322.2 | NP_001076791.1 | 7 | 514 | ||
NM_001286018.1 | NP_001272947.1 | 7 | 532 | |||
NM_033078.4 | NP_149069.1 | 7 | 607 | |||
GenBank mRNA | AF030313.1 | - | 7 | 491 | ||
AF039026.1 | - | 7 | 521 | |||
AF054819.1 | - | 8 | 781 | |||
AK040436.1 | - | 7 | 596 | |||
AK170499.1 | - | 7 | 533 | |||
AY860972.1 | - | 7 | 497 | |||
BC057147.1 | - | 7 | 558 | 5328432 | ||
CT010384.1 | - | 7 | 497 |