Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083322.2 | NP_001076791.1 | 8 | 657 | ||
NM_001286018.1 | NP_001272947.1 | 8 | 675 | |||
NM_033078.4 | NP_149069.1 | 8 | 750 | |||
GenBank mRNA | AF030313.1 | - | 8 | 634 | ||
AF039026.1 | - | 8 | 664 | |||
AF054819.1 | - | 9 | 924 | |||
AK040436.1 | - | 8 | 739 | |||
AK170499.1 | - | 8 | 676 | |||
AY860972.1 | - | 8 | 640 | |||
BC057147.1 | - | 8 | 701 | 5328432 | ||
CT010384.1 | - | 8 | 640 |