Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033561.1 | NP_291039.1 | 2 | 167 | ||
GenBank mRNA | AK045004.1 | - | 2 | 169 | ||
AK088818.1 | - | 2 | 179 | |||
AK122194.1 | - | 2 | 167 | |||
AK132034.1 | - | 2 | 187 | |||
AK135390.1 | - | 2 | 168 | |||
AK136999.1 | - | 2 | 154 | |||
AK149841.1 | - | 2 | 153 | |||
AK150088.1 | - | 2 | 176 | |||
AK166567.1 | - | 2 | 168 | |||
AK168872.1 | - | 2 | 186 | |||
AK169312.1 | - | 2 | 150 | |||
AK169353.1 | - | 2 | 168 | |||
AK169407.1 | - | 2 | 186 | |||
BC014796.1 | - | 2 | 181 | 3962104 |