Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc45a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053077.3 | NP_444307.1 | 1, 2 | 489 | ||
GenBank mRNA | AF360357.1 | - | 1, 2 | 397 | ||
AK029155.1 | - | 1, 2 | 490 | |||
AK089932.1 | - | 1, 2 | 460 | |||
AK148207.1 | - | 486 | ||||
AY034377.1 | - | 1, 2 | 410 | |||
BC125376.1 | - | 1, 2 | 476 | 40129891 | ||
BC132429.1 | - | 1, 2 | 476 | 40130706 |