Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc45a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053077.3 | NP_444307.1 | 2 | 515 | ||
GenBank mRNA | AF360357.1 | - | 2 | 423 | ||
AK029155.1 | - | 2 | 516 | |||
AK089932.1 | - | 2 | 486 | |||
AK148207.1 | - | 512 | ||||
AY034377.1 | - | 2 | 436 | |||
BC125376.1 | - | 2 | 502 | 40129891 | ||
BC132429.1 | - | 2 | 502 | 40130706 |