Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133977.2 | NP_598738.1 | 11 | 1373 | ||
GenBank mRNA | AF440692.1 | - | 10 | 1303 | ||
AK085754.1 | - | 11 | 1358 | |||
AK142599.1 | - | 1363 | ||||
AK146549.1 | - | 11 | 1358 | |||
AK149559.1 | - | 11 | 1358 | |||
AK149595.1 | - | 11 | 1358 | |||
AK150782.1 | - | 11 | 1406 | |||
AK168368.1 | - | 11 | 1360 | |||
AK168405.1 | - | 11 | 1360 | |||
AK168419.1 | - | 11 | 1358 | |||
AK168938.1 | - | 11 | 1358 | |||
AK169187.1 | - | 11 | 1357 | |||
BC008559.1 | - | 9 | 1095 | |||
BC012313.1 | - | 11 | 1373 | 3981648 | ||
BC018573.1 | - | 6 | 643 | |||
BC020295.1 | - | 11 | 1342 | |||
BC022986.1 | - | 11 | 1351 | 5123741 | ||
BC058216.1 | - | 11 | 1339 | |||
BC058218.1 | - | 11 | 1352 | |||
BC092046.1 | - | 11 | 1334 | 3594705 |