Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130457.1 | NP_001123929.1 | 3 | 413 | ||
NM_134011.2 | NP_598772.1 | 3 | 413 | |||
GenBank mRNA | AK028966.1 | - | 3 | 391 | ||
AK129245.1 | - | 3 | 368 | |||
AK144813.1 | - | 3 | 411 | |||
AK166760.1 | - | 3 | 411 | |||
AK168088.1 | - | 3 | 411 | |||
AK188017.1 | - | 1 | 28 | |||
BC016483.1 | - | 3 | 369 | 3155271 | ||
BC031155.1 | - | 3 | 384 | 4976690 | ||
BC031910.1 | - | 3 | 383 | 5362943 |