Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152229.2 | NP_689415.1 | 9 | 2374 | ||
GenBank mRNA | AK132201.1 | - | 9 | 2378 | ||
BC056950.1 | - | 9 | 2003 | |||
BC057104.1 | - | 9 | 2418 | 6401770 |