Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdhx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175094.5 | NP_780303.1 | 2 | 279 | ||
GenBank mRNA | AK046020.1 | - | 2 | 228 | ||
AK047670.1 | - | 2 | 277 | |||
AK050433.1 | - | 2 | 280 | |||
AK083170.1 | - | 2 | 243 | |||
BC061231.1 | - | 2 | 240 | 6532760 |