Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdhx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175094.5 | NP_780303.1 | 8, 9 | 1081 | ||
GenBank mRNA | AK047670.1 | - | 8, 9 | 1079 | ||
AK050433.1 | - | 8, 9 | 1082 | |||
AK083170.1 | - | 8, 9 | 1045 | |||
BC061231.1 | - | 8, 9 | 1042 | 6532760 |