Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upk1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178924.4 | NP_849255.2 | 4 | 411 | ||
GenBank mRNA | AF274865.1 | - | 3 | 346 | ||
AK053004.1 | - | 4 | 412 | |||
AK083101.1 | - | 4 | 411 | |||
AK086926.1 | - | 4 | 410 | |||
BC046604.1 | - | 4 | 389 | 6389837 |