Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upk1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178924.4 | NP_849255.2 | 8 | 850 | ||
GenBank mRNA | AF274865.1 | - | 7 | 785 | ||
AK053004.1 | - | 8 | 851 | |||
AK083101.1 | - | 8 | 850 | |||
AK086926.1 | - | 8 | 849 | |||
BC046604.1 | - | 8 | 828 | 6389837 |