Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135217.1 | NP_001128689.1 | 3 | 562 | ||
NM_006992.3 | NP_008923.1 | 3 | 390 | |||
NM_201650.2 | NP_964013.1 | 3 | 390 | |||
GenBank mRNA | AK223535.1 | - | 3 | 324 | ||
AK292291.1 | - | 3 | 325 | |||
AK292367.1 | - | 3 | 390 | |||
AY834277.1 | - | 2 | 170 | |||
BC029858.1 | - | 3 | 363 | 5170643 | ||
BM822373.1 | - | 3 | 334 | |||
U72508.1 | - | 3 | 282 |