Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136096.1 | NP_001129568.1 | 234 | |||
NM_019227.1 | NP_062100.1 | 3 | 213 | |||
GenBank mRNA | AF127798.1 | - | 3 | 213 | ||
AF132672.1 | - | 3 | 213 |