Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkar2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019264.2 | NP_062137.1 | 8 | 916 | ||
XM_006243712.2 | XP_006243774.1 | 9 | 1101 | |||
XM_006243713.2 | XP_006243775.1 | 9 | 915 | |||
XM_008766478.1 | XP_008764700.1 | 8 | 866 | |||
GenBank mRNA | AF533978.1 | - | 8 | 920 | ||
J02934.1 | - | 8 | 714 |