Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020081.1 | NP_064466.1 | 4 | 682 | ||
XM_006248399.2 | XP_006248461.1 | 4 | 1049 | |||
XM_006248400.1 | XP_006248462.1 | 3 | 658 | |||
GenBank mRNA | D50558.1 | - | 4 | 682 |