Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020306.2 | NP_064702.1 | 2 | 349 | ||
GenBank mRNA | AJ012603.1 | - | 2 | 351 |