Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020308.1 | NP_064704.1 | 6 | 544 | ||
XM_006232742.2 | XP_006232804.1 | 6 | 566 | |||
XM_006232744.2 | XP_006232806.1 | 6 | 566 | |||
XM_008761203.1 | XP_008759425.1 | 6 | 566 | |||
XM_008761204.1 | XP_008759426.1 | 6 | 566 | |||
XM_008761205.1 | XP_008759427.1 | 6 | 566 | |||
GenBank mRNA | AJ251198.1 | - | 6 | 544 | ||
BC061796.1 | - | 6 | 494 | 5597908 |