Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021744.1 | NP_068512.1 | 2 | 617 | ||
XM_006254603.2 | XP_006254665.1 | 2 | 949 | |||
GenBank mRNA | AF087943.1 | - | 2 | 617 | ||
BC061733.1 | - | 2 | 755 | 5598794 | ||
FQ219480.1 | - | 2 | 650 | |||
U51804.1 | - | 2 | 636 |