Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LECT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011705.1 | NP_001011705.1 | 4 | 501 | ||
NM_007015.2 | NP_008946.1 | 4 | 501 | |||
XM_011534897.2 | XP_011533199.1 | 4 | 499 | |||
XM_011534898.2 | XP_011533200.1 | 4 | 500 | |||
GenBank mRNA | AB006000.1 | - | 4 | 390 | ||
BC025659.1 | - | 4 | 501 | 5198878 | ||
CR541910.1 | - | 4 | 390 |