Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021849.1 | NP_068621.1 | 7 | 884 | ||
XM_006247910.2 | XP_006247972.1 | 7 | 950 | |||
XM_008768480.1 | XP_008766702.1 | 7 | 1007 | |||
XM_008768481.1 | XP_008766703.1 | 7 | 1007 | |||
GenBank mRNA | AB016486.1 | - | 7 | 884 | ||
BC062031.1 | - | 7 | 929 | 5623611 |