Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Top2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022183.2 | NP_071519.2 | 5 | 501 | ||
GenBank mRNA | Z19552.1 | - | 5 | 501 |