Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022206.1 | NP_071542.1 | 2 | 164 | ||
XM_008771505.1 | XP_008769727.1 | 2 | 403 | |||
GenBank mRNA | U87305.1 | - | 2 | 164 |