Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022230.2 | NP_071566.1 | 4 | 862 | ||
GenBank mRNA | AB030707.1 | - | 4 | 862 |