Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022246.1 | NP_071582.1 | 1, 2 | 294 | ||
GenBank mRNA | AF218576.1 | - | 1, 2 | 294 |