Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps33b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022286.2 | NP_071622.1 | 2 | 459 | ||
GenBank mRNA | BC081707.1 | - | 2 | 459 | 7128590 | |
U35245.1 | - | 2 | 174 |