Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuba1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022298.1 | NP_071634.1 | 4 | 929 | ||
GenBank mRNA | BC062238.1 | - | 4 | 963 | 6922126 | |
BC078830.1 | - | 4 | 942 | 7115494 | ||
FQ210110.1 | - | 4 | 961 | |||
FQ212756.1 | - | 4 | 896 | |||
FQ213186.1 | - | 4 | 962 | |||
FQ214446.1 | - | 2 | 518 | |||
FQ216135.1 | - | 1 | 376 | |||
FQ216180.1 | - | 1 | 376 | |||
V01227.1 | - | 4 | 929 |
Gene Symbol: Tuba1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044270.2 | NP_001037735.1 | 4 | 962 | ||
GenBank mRNA | BC060572.1 | - | 4 | 966 | 6920989 | |
BC076379.1 | - | 4 | 948 | 7106502 | ||
FQ212426.1 | - | 4 | 957 | |||
FQ213439.1 | - | 4 | 959 | |||
FQ213947.1 | - | 4 | 958 | |||
FQ214075.1 | - | 4 | 960 | |||
FQ214100.1 | - | 4 | 957 | |||
FQ214301.1 | - | 4 | 961 | |||
FQ223081.1 | - | 4 | 962 | |||
FQ225046.1 | - | 4 | 966 | |||
FQ225339.1 | - | 4 | 956 | |||
FQ225425.1 | - | 4 | 962 | |||
FQ225650.1 | - | 4 | 959 | |||
FQ231550.1 | - | 4 | 958 | |||
FQ232160.1 | - | 3 | 867 | |||
FQ232527.1 | - | 4 | 962 | |||
FQ233000.1 | - | 4 | 959 | |||
FQ233566.1 | - | 4 | 960 | |||
FQ234135.1 | - | 4 | 957 |