Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177701.2 | NP_001171172.1 | 5, 6 | 786 | ||
NM_006860.4 | NP_006851.1 | 5, 6 | 783 | |||
XM_006724106.2 | XP_006724169.1 | 6, 7 | 547 | |||
XM_017028540.1 | XP_016884029.1 | 5, 6 | 790 | |||
GenBank mRNA | AL022729.1 | - | 5, 6 | 727 | ||
BC000566.2 | - | 5, 6 | 764 | 3163341 | ||
BM711121.1 | - | 5, 6 | 374 | |||
BT006815.1 | - | 5, 6 | 360 | |||
CR456558.1 | - | 5, 6 | 402 |