Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPLP0 SIRNA Assays ›
Gene Symbol:
Rplp0
RGD1564469
Gene Name:
ribosomal protein, large, P0
similar to Acidic ribosomal phosphoprotein P0
Gene Aliases:
Arbp
-
Chromosome Location:
Chr.12: 46791528 - 46794797 on Build Rnor_6.0
Chromosome Location:
Chr.2: 206902779 - 206903920 on Build Rnor_6.0
UniGene ID:
Rn.1079
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID
189378
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rplp0
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022402.2
NP_071797.1
3
368
GenBank mRNA
BC062028.1
-
2
337
5623881
DN931898.1
-
2
386
FQ209537.1
-
3
368
FQ209603.1
-
3
368
FQ209620.1
-
2
353
FQ209649.1
-
3
368
FQ209757.1
-
3
369
FQ210412.1
-
3
368
FQ211152.1
-
3
368
FQ211263.1
-
3
367
FQ212554.1
-
3
368
FQ212743.1
-
3
368
FQ212875.1
-
3
368
FQ212927.1
-
3
369
FQ213085.1
-
3
368
FQ213427.1
-
3
368
FQ213510.1
-
3
368
FQ213712.1
-
3
368
FQ213847.1
-
3
369
FQ214087.1
-
3
368
FQ214354.1
-
3
368
FQ214515.1
-
3
368
FQ214553.1
-
3
369
FQ214850.1
-
3
368
FQ214997.1
-
3
368
FQ215007.1
-
3
368
FQ215548.1
-
3
370
FQ215662.1
-
3
368
FQ215682.1
-
3
368
FQ215870.1
-
3
368
FQ216120.1
-
3
369
FQ216296.1
-
3
368
FQ216389.1
-
3
369
FQ216406.1
-
3
371
FQ216495.1
-
3
368
FQ216618.1
-
3
368
FQ216655.1
-
3
369
FQ216733.1
-
3
370
FQ216774.1
-
3
368
FQ216813.1
-
3
367
FQ216851.1
-
3
368
FQ216913.1
-
3
369
FQ217406.1
-
3
368
FQ218561.1
-
3
368
FQ218832.1
-
3
368
FQ218943.1
-
3
368
FQ219025.1
-
3
371
FQ219184.1
-
3
372
FQ219389.1
-
3
368
FQ219457.1
-
3
368
FQ219678.1
-
3
368
FQ219815.1
-
3
368
FQ219817.1
-
3
368
FQ219837.1
-
3
368
FQ219855.1
-
3
368
FQ219923.1
-
3
368
FQ220008.1
-
3
368
FQ220051.1
-
3
368
FQ220077.1
-
3
368
FQ220115.1
-
3
368
FQ220181.1
-
3
368
FQ220201.1
-
3
366
FQ220213.1
-
3
368
FQ220247.1
-
3
369
FQ220282.1
-
3
370
FQ220284.1
-
3
368
FQ220302.1
-
3
368
FQ220338.1
-
3
368
FQ220345.1
-
3
368
FQ220370.1
-
3
368
FQ220406.1
-
3
368
FQ220413.1
-
3
368
FQ220420.1
-
3
369
FQ220433.1
-
3
365
FQ220444.1
-
3
368
FQ220445.1
-
3
364
FQ220449.1
-
3
368
FQ220500.1
-
3
368
FQ220514.1
-
3
368
FQ220527.1
-
3
368
FQ220533.1
-
3
400
FQ220542.1
-
3
368
FQ220545.1
-
3
368
FQ220634.1
-
3
368
FQ220654.1
-
3
368
FQ220684.1
-
3
369
FQ220690.1
-
3
370
FQ220769.1
-
3
368
FQ220801.1
-
3
368
FQ220833.1
-
3
368
FQ220838.1
-
3
368
FQ220888.1
-
3
368
FQ220932.1
-
3
369
FQ221146.1
-
3
368
FQ221380.1
-
3
368
FQ221533.1
-
3
370
FQ221556.1
-
3
366
FQ222171.1
-
3
368
FQ222181.1
-
3
368
FQ222327.1
-
3
368
FQ222772.1
-
3
368
FQ222906.1
-
3
368
FQ223121.1
-
3
368
FQ223487.1
-
3
368
FQ224547.1
-
3
368
FQ224834.1
-
3
368
FQ224846.1
-
3
368
FQ225049.1
-
3
368
FQ225170.1
-
3
368
FQ225608.1
-
3
368
FQ225654.1
-
3
370
FQ225860.1
-
3
370
FQ225886.1
-
3
368
FQ226306.1
-
3
370
FQ227381.1
-
3
368
FQ227522.1
-
3
363
FQ227719.1
-
3
368
FQ227837.1
-
3
368
FQ227937.1
-
3
368
FQ227941.1
-
3
368
FQ227942.1
-
3
488
FQ228548.1
-
3
368
FQ228995.1
-
3
368
FQ228996.1
-
3
368
FQ229139.1
-
3
370
FQ229159.1
-
3
368
FQ229164.1
-
3
369
FQ229208.1
-
3
368
FQ229241.1
-
3
368
FQ229244.1
-
3
368
FQ229248.1
-
1
248
FQ229272.1
-
3
368
FQ229273.1
-
3
369
FQ229303.1
-
3
368
FQ229337.1
-
3
368
FQ229352.1
-
3
368
FQ229365.1
-
3
368
FQ229397.1
-
3
368
FQ229408.1
-
3
369
FQ229433.1
-
3
368
FQ229448.1
-
3
368
FQ229521.1
-
3
368
FQ229547.1
-
3
368
FQ229554.1
-
3
368
FQ229566.1
-
3
364
FQ229610.1
-
3
368
FQ229626.1
-
3
365
FQ229726.1
-
3
368
FQ229740.1
-
3
368
FQ229742.1
-
3
368
FQ229776.1
-
3
368
FQ229808.1
-
3
368
FQ229815.1
-
3
368
FQ229822.1
-
3
368
FQ229830.1
-
3
370
FQ229883.1
-
3
370
FQ229901.1
-
3
368
FQ229932.1
-
1
266
FQ229952.1
-
3
368
FQ229974.1
-
3
368
FQ230005.1
-
3
368
FQ230055.1
-
3
367
FQ230127.1
-
3
369
FQ230267.1
-
3
369
FQ230943.1
-
3
368
FQ231445.1
-
3
370
FQ231525.1
-
3
368
FQ231591.1
-
3
370
FQ231710.1
-
3
369
FQ231733.1
-
3
369
FQ231897.1
-
3
370
FQ231967.1
-
3
368
FQ232029.1
-
3
369
FQ232154.1
-
3
368
FQ232165.1
-
3
368
FQ232310.1
-
3
367
FQ232319.1
-
3
369
FQ232767.1
-
3
368
FQ232951.1
-
3
368
FQ233152.1
-
3
367
FQ233377.1
-
3
368
FQ233473.1
-
3
368
FQ233732.1
-
3
368
FQ233737.1
-
3
368
FQ233808.1
-
3
368
FQ234195.1
-
3
368
FQ234613.1
-
3
368
FQ234733.1
-
3
368
FQ234863.1
-
3
368
FQ235063.1
-
3
368
FQ235079.1
-
3
368
FQ235122.1
-
3
368
FQ235259.1
-
3
368
X15096.1
-
3
368
Z29530.1
-
2
324
Gene Symbol: RGD1564469
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_227546.7
XP_227546.4
2
367
Back To Top
More Information
Panther Classification:
Molecular Function -
ribosomal protein
translational protein
Gene Ontology Categories:
Function(s)
Process(es)
translational elongation
ribosome biogenesis
cellular response to interleukin-4
structural constituent of ribosome
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline