Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022509.1 | NP_071954.1 | 3 | 208 | ||
GenBank mRNA | AF044910.1 | - | 3 | 188 | ||
AY876898.1 | - | 3 | 178 | |||
BC062404.1 | - | 3 | 193 | 5621519 | ||
FQ212707.1 | - | 3 | 204 | |||
U75369.1 | - | 3 | 208 |