Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022547.1 | NP_071992.1 | 3 | 652 | ||
GenBank mRNA | BC089101.1 | - | 3 | 661 | 7306458 | |
M59861.1 | - | 3 | 652 |