Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284308.1 | NP_001271237.1 | 9 | 883 | ||
NM_001284309.1 | NP_001271238.1 | 9 | 823 | |||
NM_001284310.1 | NP_001271239.1 | 8 | 798 | |||
NM_001284311.1 | NP_001271240.1 | 7 | 651 | |||
NM_006869.3 | NP_006860.1 | 9 | 1056 | |||
GenBank mRNA | AB097049.1 | - | 9 | 1030 | ||
AF082324.1 | - | 9 | 1030 | |||
AJ006422.1 | - | 9 | 1052 | |||
AK092471.1 | - | 8 | 800 | |||
AK124979.1 | - | 7 | 902 | |||
AK290517.1 | - | 10 | 1002 | |||
AK300644.1 | - | 7 | 578 | |||
AK300859.1 | - | 7 | 638 | |||
AK300999.1 | - | 9 | 883 | |||
BC033747.1 | - | 9 | 857 | 5195416 |