Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022692.1 | NP_073183.1 | 185 | |||
GenBank mRNA | AF072935.1 | - | 185 | |||
BC161848.1 | - | 511 | 9025210 |
Gene Symbol: Rab5al1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751377.3 | XP_003751425.1 | 1 | 417 |